Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hog-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270099.3 | NP_001257028.1 | 3 - 4 | 323 | 67 | |
NM_001270100.3 | NP_001257029.1 | 1 - 2 | 84 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |