Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076593.8 | NP_508994.1 | 3 - 4 | 437 | 109 | |
GenBank mRNA | AB126261.1 | - | 3 - 4 | 417 | 109 |
Gene Symbol: acs-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076592.9 | NP_508993.2 | 3 - 4 | 432 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |