Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mom-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076653.5 | NP_509054.1 | 1 - 2 | 146 | 114 | |
GenBank mRNA | AF013489.1 | - | 1 - 2 | 143 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |