Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381009.1 | NP_001368711.1 | 6 - 7 | 733 | 84 | |
NM_001381010.2 | NP_001367505.1 | 6 - 7 | 1029 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |