Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076727.9 | NP_509128.1 | 5 - 6 | 857 | 91 | |
GenBank mRNA | X86403.1 | - | 736 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |