Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sur-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076828.8 | NP_509229.1 | 4 - 5 | 512 | 78 | |
GenBank mRNA | AY091467.1 | - | 3 - 4 | 443 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |