Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mek-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029600.6 | NP_001024771.1 | 2 - 3 | 526 | 91 | |
NR_132545.1 | 1 - 2 | 181 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |