Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076869.6 | NP_509270.1 | 8 - 9 | 1056 | 129 | |
GenBank mRNA | AY519852.1 | - | 8 - 9 | 1056 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |