Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076883.7 | NP_509284.2 | 2 - 3 | 286 | 65 | |
GenBank mRNA | AF515600.1 | - | 2 - 3 | 211 | 65 |
Gene Symbol: apy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076882.10 | NP_509283.1 | 2 - 3 | 160 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |