Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kvs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029318.5 | NP_001024489.1 | 11 - 12 | 1462 | 140 | |
NM_001029319.4 | NP_001024490.1 | 11 - 12 | 1436 | 140 | ||
NM_001373368.2 | NP_001360715.1 | 12 - 13 | 1493 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |