Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ksr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076995.7 | NP_509396.1 | 11 - 12 | 1944 | 100 | |
GenBank mRNA | S80647.1 | - | 11 - 12 | 1965 | 100 | |
U38820.1 | - | 11 - 12 | 1944 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |