Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077023.5 | NP_509424.2 | 6 - 7 | 876 | 137 | |
GenBank mRNA | AY887909.1 | - | 6 - 7 | 876 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |