Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077026.10 | NP_509427.2 | 1 - 2 | 169 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |