Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077037.7 | NP_509438.1 | 15 - 16 | 1997 | 137 | |
GenBank mRNA | L25312.1 | - | 15 - 16 | 1939 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |