Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077037.7 | NP_509438.1 | 5 - 6 | 945 | 63 | |
GenBank mRNA | L25312.1 | - | 5 - 6 | 887 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |