Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077156.6 | NP_509557.1 | 4 - 5 | 390 | 140 | |
GenBank mRNA | AY008134.1 | - | 3 - 4 | 318 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |