Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270149.4 | NP_001257078.1 | 3 - 4 | 348 | 120 | |
NM_001270150.3 | NP_001257079.1 | 3 - 4 | 351 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |