Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: him-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029411.3 | NP_001024582.1 | 10 - 11 | 3215 | 74 | |
NM_001373449.3 | NP_001359724.1 | 10 | 3154 | 74 | ||
NM_001373450.2 | NP_001360633.1 | 9 - 10 | 2860 | 74 | ||
NM_001373451.2 | NP_001360634.1 | 9 - 10 | 2860 | 74 | ||
NM_077234.7 | NP_509635.2 | 10 - 11 | 3154 | 74 | ||
GenBank mRNA | AF074901.1 | - | 10 - 11 | 3154 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |