Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129644.2 | NP_001123116.1 | 9 - 10 | 933 | 102 | |
NM_001373473.2 | NP_001360683.1 | 9 - 10 | 933 | 102 | ||
NM_001373474.1 | NP_001359532.1 | 9 - 10 | 933 | 102 | ||
NM_001373475.2 | NP_001360682.1 | 9 - 10 | 934 | 102 | ||
NM_001373476.2 | NP_001360686.1 | 9 - 10 | 933 | 102 | ||
NM_001373477.1 | NP_001360687.1 | 9 - 10 | 933 | 102 | ||
NM_001373478.3 | NP_001360684.1 | 10 | 976 | 102 | ||
NM_001373479.2 | NP_001360685.1 | 10 | 981 | 102 | ||
NM_001373480.3 | NP_001360688.1 | 10 | 982 | 102 | ||
NM_001373481.2 | NP_001360689.1 | 10 | 972 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |