Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029376.6 | NP_001024547.1 | 15 - 16 | 2088 | 99 | |
NM_001047774.5 | NP_001041239.1 | 14 - 15 | 1986 | 99 | ||
NM_001381088.1 | NP_001366986.1 | 13 - 14 | 1911 | 99 | ||
NM_001381089.2 | NP_001366987.1 | 3 - 4 | 630 | 99 | ||
GenBank mRNA | AF136238.1 | - | 15 - 16 | 2121 | 99 | |
AF136239.1 | - | 13 - 14 | 1918 | 99 | ||
AF136240.1 | - | 3 - 4 | 630 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |