Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373491.2 | NP_001360639.1 | 21 - 22 | 2767 | 91 | |
NM_001373492.2 | NP_001360635.1 | 21 - 22 | 2767 | 91 | ||
NM_001373495.2 | NP_001360638.1 | 22 - 23 | 2938 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |