Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slo-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029356.4 | NP_001024527.1 | 18 - 19 | 3521 | 120 | |
NM_001029357.5 | NP_001024528.1 | 17 - 18 | 3485 | 120 | ||
NM_001029358.5 | NP_001024529.1 | 17 - 18 | 3166 | 120 | ||
NM_001136452.4 | NP_001129924.1 | 18 - 19 | 3205 | 120 | ||
NM_001270215.3 | NP_001257144.1 | 19 - 20 | 3277 | 120 | ||
NM_001270216.3 | NP_001257145.1 | 18 - 19 | 3232 | 120 | ||
NM_001270217.3 | NP_001257146.1 | 18 - 19 | 3286 | 120 | ||
NM_001270218.3 | NP_001257147.1 | 18 - 19 | 3247 | 120 | ||
GenBank mRNA | AF173828.1 | - | 18 - 19 | 3413 | 120 | |
AF232770.1 | - | 17 - 18 | 3178 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |