Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077535.7 | NP_509936.3 | 16 - 17 | 1987 | 71 | |
GenBank mRNA | AY887904.1 | - | 16 - 17 | 2009 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |