Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sto-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077540.2 | NP_509941.1 | 2 - 3 | 346 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |