Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cab-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077635.6 | NP_510036.1 | 5 - 6 | 801 | 63 | |
GenBank mRNA | AF293976.1 | - | 3 - 4 | 614 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |