Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077667.1 | NP_510068.1 | 3 - 4 | 622 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |