Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077861.6 | NP_510262.1 | 1 - 2 | 345 | 84 | |
GenBank mRNA | AF187013.1 | - | 1 - 2 | 296 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |