Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077861.6 | NP_510262.1 | 2 - 3 | 457 | 65 | |
GenBank mRNA | AF187013.1 | - | 2 - 3 | 408 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |