Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077866.4 | NP_510267.2 | 1 - 2 | 100 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |