Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373569.1 | NP_001359773.1 | 2 - 3 | 596 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |