Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ztf-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373576.2 | NP_001360586.1 | 3 - 4 | 643 | 86 | |
NM_001373577.2 | NP_001360587.1 | 3 | 608 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |