Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078018.2 | NP_510419.1 | 2 - 3 | 170 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |