Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxb-76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069745.2 | NP_502146.1 | 1 - 2 | 72 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |