Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fce-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028776.3 | NP_001023947.1 | 2 - 3 | 196 | 69 | |
GenBank mRNA | AJ487543.1 | - | 2 - 3 | 218 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |