Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028964.4 | NP_001024135.2 | 2 - 3 | 501 | 171 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |