Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028978.3 | NP_001024149.1 | 3 - 4 | 506 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |