Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hil-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026111.5 | NP_001021282.1 | 1 | 88 | 61 | |
NM_001026112.4 | NP_001021283.1 | 4 | 1411 | 61 | ||
NM_001380286.1 | NP_001367386.1 | 1 | 48 | 61 | ||
NM_068626.8 | NP_501027.3 | 1 | 428 | 61 | ||
GenBank mRNA | AF216291.1 | - | 1 | 80 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Both primers and probe map within a single exon |