Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srz-82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026144.1 | NP_001021315.1 | 3 - 4 | 559 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |