Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026882.3 | NP_001022053.1 | 3 - 4 | 419 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |