Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038424.4 | NP_001033513.2 | 6 - 7 | 585 | 141 | |
NM_001330830.1 | NP_001317792.1 | 6 - 7 | 539 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |