Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl-7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171316.8 | NP_741371.2 | 4 | 669 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Amplicon spans exons and probe does not span exons |