Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eat-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026522.3 | NP_001021693.1 | 1 | 77 | 59 | |
NM_001383759.1 | 1 | 136 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Both primers and probe map within a single exon |