Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttr-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047494.2 | NP_001040959.1 | 2 - 3 | 292 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |