Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628193.3 | XP_005628250.1 | 4 - 5 | 2499 | 114 | |
XM_022426995.1 | XP_022282703.1 | 4 - 5 | 362 | 114 | ||
XM_022426996.1 | XP_022282704.1 | 5 - 6 | 2693 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |