Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538650.6 | XP_538650.2 | 1 - 2 | 803 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |