Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003072.1 | NP_001003072.1 | 1 - 2 | 966 | 72 | |
XM_005634674.3 | XP_005634731.1 | 2 - 3 | 1491 | 72 | ||
XM_005634675.3 | XP_005634732.1 | 3 - 4 | 1008 | 72 | ||
XM_005634676.3 | XP_005634733.1 | 2 - 3 | 1509 | 72 | ||
XM_005634677.3 | XP_005634734.1 | 3 - 4 | 1122 | 72 | ||
XM_014106833.2 | XP_013962308.1 | 3 - 4 | 1140 | 72 | ||
XM_014106834.2 | XP_013962309.1 | 3 - 4 | 1026 | 72 | ||
GenBank mRNA | AB073983.1 | - | 1 - 2 | 966 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Inventoried Probe spans exons |