Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003007.2 | NP_001003007.2 | 7 - 8 | 814 | 69 | |
GenBank mRNA | AB100595.1 | - | 6 - 7 | 768 | 69 | |
DN325679.1 | - | 1 - 2 | 115 | 69 | ||
DN359992.1 | - | 2 - 3 | 234 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |