Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005252.1 | NP_001005252.1 | 1 - 2 | 76 | 86 | |
XM_022422563.1 | XP_022278271.1 | 2 - 3 | 360 | 86 | ||
GenBank mRNA | AB163918.1 | - | 1 - 2 | 76 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons |