Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003339.1 | NP_001003339.1 | 2 - 3 | 342 | 65 | |
GenBank mRNA | AF029978.1 | - | 2 - 3 | 340 | 65 | |
AF029979.1 | - | 2 - 3 | 342 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |