Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197044.1 | NP_001183973.1 | 3 - 4 | 496 | 70 | |
GenBank mRNA | AF049909.1 | - | 3 - 4 | 477 | 70 | |
HG965164.1 | - | 3 - 4 | 485 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |